Skip to Content
Merck
All Photos(1)

Key Documents

T75698

Sigma-Aldrich

2,3,3-Trimethyl-1-butene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC(CH3)=CH2
CAS Number:
Molecular Weight:
98.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.39 (vs air)

vapor pressure

190 mmHg ( 37.7 °C)

Assay

98%

autoignition temp.

707 °F

refractive index

n20/D 1.402 (lit.)

bp

78-80 °C (lit.)

density

0.705 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(C)(C)C

InChI

1S/C7H14/c1-6(2)7(3,4)5/h1H2,2-5H3

InChI key

AUYRUAVCWOAHQN-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service