Skip to Content
Merck
All Photos(1)

Documents

T62006

Sigma-Aldrich

Trifluoroacetaldehyde ethyl hemiacetal

90%

Synonym(s):

1-Ethoxy-2,2,2-trifluoroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH(OH)OCH2CH3
CAS Number:
Molecular Weight:
144.09
Beilstein:
906797
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

impurities

≤12% water

refractive index

n20/D 1.342 (lit.)

bp

104-105 °C/745 mmHg (lit.)

density

1.221 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[Na+].[O-]c1ccccc1

InChI

1S/C4H7F3O2/c1-2-9-3(8)4(5,6)7/h3,8H,2H2,1H3

InChI key

KLXJPQNHFFMLIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Alternative to gaseous unstable trifluoroacetaldehyde: used to prepare α-trifluoromethylated alcohols for antifungals, antitumor, and chemotherapeutic agents.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 2, 235-235 (1991)
Tetrahedron Letters, 32, 3823-3823 (1991)
Tetrahedron Letters, 33, 1351-1351 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service