Skip to Content
Merck
All Photos(1)

Key Documents

T31704

Sigma-Aldrich

N-Thionylaniline

98%

Synonym(s):

N-Sulfinyl-aniline

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$178.00

$178.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$178.00

About This Item

Linear Formula:
C6H5NSO
CAS Number:
Molecular Weight:
139.18
Beilstein:
507682
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$178.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.627 (lit.)

bp

200 °C (lit.)
83 °C/15 mmHg (lit.)

density

1.236 g/mL at 25 °C (lit.)

SMILES string

O=S=Nc1ccccc1

InChI

1S/C6H5NOS/c8-9-7-6-4-2-1-3-5-6/h1-5H

InChI key

FIOJWGRGPONADF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile synthetic intermediate.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ary R Murad et al.
Polymers, 13(1) (2020-12-31)
Two novel low band gap donor-acceptor (D-A) copolymers, poly[9,10-bis(4-(dodecyloxy)phenyl)-2,6-anthracene-alt-5,5-(4',7'-bis(2-thienyl)-2',1',3'-benzothiadiazole-N-5,6-(3,7-dimethyloctyl)dicarboxylic imide)] (PPADTBTDI-DMO) and poly[9,10-bis(4-(dodecyloxy)phenyl)-2,6-anthracene-alt-5,5-(4',7'-bis(2-thienyl)-2',1',3'-benzothiadiazole-5,6-N-octyl-dicarboxylic imide)] (PPADTBTDI-8) were synthesized in the present work by copolymerising the bis-boronate ester of 9,10-phenylsubstituted anthracene flanked by thienyl groups as electron-donor units with benzothiadiazole dicarboxylic imide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service