Key Documents
SYNS740100
Synthware sublimation apparatus, complete
100 mL, complete
Synonym(s):
Synthware S740100, sublimator
Select a Size
1 EA
$739.00
$739.00
Check Cart for Availability
Select a Size
Change View
1 EA
$739.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
$739.00
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S740100
capacity
100 mL
100 mL
Related Categories
General description
Two-piece grease-free sublimer with a flat bottom to enable heating on a hot plate stirrer. Condenser has a large top opening for use with dry ice/acetone or liquid nitrogen. Connection between the flask and condenser is made with a flat O-ring joint permitting parts to be gently separated for maximum product recovery. The capacity listed is that below the bottom of the cold finger. Vacuum control is provided by a 0-8mm high vacuum valve. Complete apparatus consists of the condenser, flask, Viton® O-ring
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)