Skip to Content
Merck
All Photos(1)

Key Documents

SYNP120025M

Synthware peptide synthesis vessel with fritted disc and PTFE stopcock

25 mL, disc O.D. 20 mm, porosity: medium, GL 25

Synonym(s):

Synthware P120025M, Solid phase peptide synthesis vessel

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$230.00

$230.00


Check Cart for Availability


Select a Size

Change View
1 EA
$230.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.51

$230.00


Check Cart for Availability

packaging

pkg of 1 ea

manufacturer/tradename

Synthware P120025M

capacity

25 mL
25 mL

disc O.D.

20 mm

fitting

GL 25

Looking for similar products? Visit Product Comparison Guide

General description

With a fritted disc of coarse or medium porosity to support resin. Top of the vessel has GL thread and a PTFE lined screw cap. PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service