Key Documents
SYNH101410
Synthware Hickman still head with cap, o-ring and PTFE-faced septum
joint: ST/NS 14/10
Synonym(s):
Synthware H101410, Hickman Distillation head
Select a Size
1 EA
$82.90
$82.90
Check Cart for Availability
Select a Size
Change View
1 EA
$82.90
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$82.90
Check Cart for Availability
Joint
joint: ST/NS14/10
manufacturer/tradename
H101410
Related Categories
General description
Synthware Hickman distillation head is designed for microscale fractional distillation. It is supplied with one compression cap, one Viton® O-ring and one PTFE - faced silicone rubber septum. Replacement accessories available.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)