Key Documents
SYNF984250
Synthware Kuderna-Danish flask
capacity 250 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 19/22
Synonym(s):
Synthware F984250
Select a Size
1 EA
$102.00
$102.00
Check Cart for Availability
Select a Size
Change View
1 EA
$102.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$102.00
Check Cart for Availability
material
borosilicate glass flask
pear-shaped
single-neck
Joint
bottom joint: ST/NS19/22
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F984250
capacity
250 mL
General description
Top joint is a 24/40 outer while the lower joint is a 19/22 inner.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)