Key Documents
SYNF714500
Synthware four-neck round bottom flask with threaded side arm
capacity 500 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonym(s):
Synthware F714500, 4-neck flask, multi-neck reaction flask
Select a Size
1 EA
$253.00
$253.00
Check Cart for Availability
Select a Size
Change View
1 EA
$253.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$253.00
Check Cart for Availability
material
borosilicate glass flask
four-neck (with threaded side arm)
round bottom
Joint
center joint: ST/NS24/40
side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F714500
capacity
500 mL
General description
Round bottom, four-neck flask with standard taper outer joints and a thermometer joints. Supplied complete with a compression cap and Viton® O-ring. For insertion of thermometers or bleed tubes having an O.D. between 5.0-7.0 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)