SYNF664500
Synthware™ Erlenmeyer flask
capacity 500 mL, joint: ST/NS 24/40
Synonym(s):
Synthware F664500, glass conical flask
All Photos(1)
About This Item
Recommended Products
material
flat bottom
glass flask
Joint
joint neck24/40
joint: ST/NS24/40
sterility
non-sterile
feature
without cap
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F664500
capacity
500 mL
Related Categories
General description
With joint. Stopper is not supplied. All sizes including 1L flasks are hand-blown from tubing to ensure heavy and uniform wall.
Other Notes
Browse all Erlenmeyer flasks
Legal Information
Synthware is a trademark of Kemtech America Inc.