Key Documents
SYNF642460
Synthware pressure equalizing funnel with high vacuum valve
60 mL, joint: ST/NS 24/40, valve size 0-4 mm
Synonym(s):
Synthware F642460, addition funnel
Select a Size
1 EA
$342.00
$342.00
Estimated to ship on31 May 2025
Select a Size
Change View
1 EA
$342.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
$342.00
Estimated to ship on31 May 2025
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F642460
capacity
60 mL
60 mL
Related Categories
General description
With PTFE extended tip valve for semi-needle valve adjustment.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)