Key Documents
SYNF591050
Synthware three-neck round bottom flask
50 mL, joint: ST/NS 14/10
Synonym(s):
Synthware F591050, 3-neck flask
Select a Size
1 EA
$117.00
$117.00
Check Cart for Availability
Select a Size
Change View
1 EA
$117.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$117.00
Check Cart for Availability
material
borosilicate glass flask
round bottom
three-neck
Joint
joint: ST/NS14/10
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F591050
capacity
50 mL
50 mL
General description
Microscale, round bottom flask with three 14/10 outer ground joints externally threaded. For replacement caps, see RC112; for Viton® O-rings, see R014; for septa, see R1420.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)