Key Documents
SYNF484258
Synthware solvent transfer/storage flask
250 mL, joint: ST/NS 24/40, valve size 0-8 mm
Synonym(s):
Synthware F484258, Straus flask, receiver flask
Select a Size
1 EA
$165.00
$165.00
Check Cart for Availability
Select a Size
Change View
1 EA
$165.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$165.00
Check Cart for Availability
material
borosilicate glass flask
round bottom
two-neck
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F484258
capacity
250 mL
250 mL
valve size
0-8 mm
General description
Synthware solvent transfer/storage flask features a center neck joint sealed off from direct access to flask. A syringe or cannula is used to recover the solvent through the valve.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)