Key Documents
SYNF479050
Synthware Squibb-style separatory funnel with PTFE stopcock and PTFE stopper
50 mL, top joint: ST/NS 19/22
Synonym(s):
Synthware F479050, glass separatory funnel
Select a Size
1 EA
$187.00
$187.00
Check Cart for Availability
Select a Size
Change View
1 EA
$187.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
$187.00
Check Cart for Availability
Joint
joint: ST/NS top19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F479050
capacity
50 mL
50 mL
Related Categories
General description
Supplied with a Glass or PTFE stopper
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)