Key Documents
SYNF411150
Synthware two neck round bottom flask with angled side neck
50 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonym(s):
Synthware F411150, 2-neck round flask
Select a Size
1 EA
$76.00
$76.00
Available to ship on28 April 2025Details
Select a Size
Change View
1 EA
$76.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$76.00
Available to ship on28 April 2025Details
material
borosilicate glass flask
round bottom
two-neck (angled side neck)
Joint
center joint: ST/NS14/20
side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F411150
capacity
50 mL
50 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)