Key Documents
SYNA462424
Synthware modified anti-splash adapter with fritted disc
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A462424, Anti-splash adapter, splash head, bump trap
Select a Size
1 EA
$221.00
$221.00
Check Cart for Availability
Select a Size
Change View
1 EA
$221.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$221.00
Check Cart for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A462424
capacity
100 mL
Related Categories
General description
With extra coarse fritted disc sealed above lower inner joint. Frit reduces foam overflow from being drawn into the condenser.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)