Key Documents
SYNA351410
Synthware distillation adapter
joint: ST/NS 14/10
Synonym(s):
Synthware A351410, adapter, sidearm adapter
Select a Size
1 EA
$67.10
$67.10
Check Cart for Availability
Select a Size
Change View
1 EA
$67.10
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$67.10
Check Cart for Availability
Joint
joint: ST/NS14/10
packaging
pack of 1 ea
manufacturer/tradename
A351410
Related Categories
General description
Synthware distillation side arm adapter with 14/10 joint. Supplied with one holed compression cap and one ViltonTM O-ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)