Key Documents
SYNA301922
Synthware vacuum or argon/nitrogen adapter with hose connection on top
joint: ST/NS 19/22
Synonym(s):
Synthware A301922, inert gas adapter, vacuum adapter
Select a Size
1 EA
$29.10
$29.10
Check Cart for Availability
Select a Size
Change View
1 EA
$29.10
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$29.10
Check Cart for Availability
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A301922
Related Categories
General description
With hose connection on the top and male joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)