Key Documents
SYNA301410
Synthware gas inlet/outlet adapter
joint: ST/NS 14/10
Synonym(s):
Synthware A301410, inert gas adapter, vacuum adapter
Select a Size
1 EA
$31.20
$31.20
Check Cart for Availability
Select a Size
Change View
1 EA
$31.20
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$31.20
Check Cart for Availability
Joint
joint: ST/NS14/10
packaging
pack of 1 ea
manufacturer/tradename
A301410
Related Categories
General description
Synthware gas inlet/outlet adapter is used to facilitate the introduction and removal of gases in various system. The joint is 14/10 and the hose connection is 8 mm O.D.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)