Key Documents
SYNA281250
Synthware anti-splash adapter
250 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 14/20
Synonym(s):
Synthware A281250, rotary evaporator bump trap, bump trap
Select a Size
1 EA
$155.00
$155.00
Check Cart for Availability
Select a Size
Change View
1 EA
$155.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$155.00
Check Cart for Availability
Joint
bottom joint: ST/NS14/20
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A281250
capacity
250 mL
Related Categories
General description
This design prevents possible contamination from rotary evaporator.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)