Skip to Content
Merck
All Photos(1)

Documents

SLW3700/06

Pyrex® Büchner filter funnels with sintered disc

125 mL, disc diam. 65 mm, porosity grade 3

Synonym(s):

Pyrex, SciLabware 3700/06

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

manufacturer/tradename

3700/06
SciLabware

capacity

125 mL
125 mL

disc diam.

65 mm

pore size

grade 3 porosity

Looking for similar products? Visit Product Comparison Guide

General description

The 60mm sintered discs fitted to these funnels are manufactured from the same 3.3 expansion borosilicate glass as the funnel bodies, making these funnels suitable to be used with a wide range aggressive chemicals.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service