SLW3660/02M
Pyrex® Gooch crucible
30 mL, disc diam. 30 mm, porosity grade 1
Synonym(s):
Pyrex, SciLabware 3660/02M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
manufacturer/tradename
3660/02M
SciLabware
disc diam.
30 mm
pore size
grade 1 porosity
capacity
30 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Ideal for drying precipitates to constant mass at 110 deg. C
- Also suitable for ignition work up to 450 deg. C (see safety notice)
- With integral glass sintered disc
- Manufactured from chemically resistant Pyrex borosilicate glass
Features and Benefits
• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• Sintered glass filter discs comply with ISO 4793• Ideal for drying precipitates to constant mass at 110°C• Also suitable for ignition work up to 450°C (see safety notice below)
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service