Skip to Content
Merck
All Photos(1)

Key Documents

SLW3620/08

Pyrex® Büchner filter funnels with sintered disc

30 mL, disc diam. 30 mm, porosity grade 4

Synonym(s):

Pyrex, SciLabware 3620/08

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43
Pricing and availability is not currently available.

manufacturer/tradename

3620/08
SciLabware

capacity

30 mL
30 mL

disc diam.

30 mm

pore size

grade 4 porosity

Looking for similar products? Visit Product Comparison Guide

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service