Key Documents
SLW3152/08A
Pyrex® Volumetric flask, heavy duty, class A, amber, USP/ISO/DIN tolerances,
capacity 25 mL
Synonym(s):
Pyrex, SciLabware 3152/08A
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass flask
Joint
joint ST/NS neck10/19
feature
accuracy class A (ISO 1042 and DIN 12664, United States Pharmacopoeia (USP),conformity ′H′ certified)
packaging
pkg of 5 ea
manufacturer/tradename
SciLabware 3152/08A
Overall H
110 mm
capacity
25 mL
General description
  • Complies with ISO 1042 and DIN 12664
  • Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
  • Manufactured from borosilicate glass with an amber coating on exterior surfaces to facilitate use with light sensitive solutions
  • Each flask is laser etched with an individual serial number, batch code and date of manufacturer
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)