Key Documents
SLW1132/15
Pyrex® Erlenmeyer flask, graduated, ground socket
joint: 29/32, capacity 250 mL
Synonym(s):
Pyrex, SciLabware 1132/15
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
flat bottom
Joint
joint neck29/32
joint: 29/32
description
ground joint
sterility
non-sterile
feature
graduated
without cap
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1132/15
capacity
250 mL
diam.
85 mm
General description
  • Erlenmeyer flasks with high quality Quickfit® ground glass socket
  • Manufactured from Pyrex borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
Features and Benefits
  • Complies with ISO 1773 and 4797
  • Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
  • Easy-to-read white enamel marking spot and graduated scale allows the approximate measurement of the flask contents
  • N.B. Conical shape flasks are not suitable for use under vacuum, unless specifically engineered to do so.
Other Notes
Legal Information
PYREX is a registered trademark of Corning, Inc.
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)