Key Documents
SLW1060/09D
Pyrex® Watch glasses
nominal diameter 60 mm
Synonym(s):
Pyrex, SciLabware 1060/09D
About This Item
UNSPSC Code:
41121801
NACRES:
NB.43
Pricing and availability is not currently available.
packaging
pkg of 10
manufacturer/tradename
1060/09D
SciLabware
nominal diameter
60 mm
General description
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)