Skip to Content
Merck
All Photos(1)

Key Documents

SIK4711-30

Sigma-Aldrich

11-Azidoundecyltriethoxysilane

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$961.00

$961.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$961.00

About This Item

Empirical Formula (Hill Notation):
C17H37N3O3Si
CAS Number:
Molecular Weight:
359.58
UNSPSC Code:
12352103
NACRES:
NA.23

$961.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

liquid (colorless)

shelf life

3 years under the recommended conditions

Application

"CLICK CHEMISTRY"
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

Legal Information

Product of SiKEMIA

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jieming Li et al.
Nature nanotechnology, 13(8), 723-729 (2018-05-08)
Dynamic DNA nanotechnology has yielded nontrivial autonomous behaviours such as stimulus-guided locomotion, computation and programmable molecular assembly. Despite these successes, DNA-based nanomachines suffer from slow kinetics, requiring several minutes or longer to carry out a handful of operations. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service