Skip to Content
Merck
All Photos(1)

Key Documents

SIK2701-10

Sigma-Aldrich

3,3′,5,5′-Azobenzenetetracarboxylic acid - TAZBH4

≥97%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$150.00
1 G
$436.00

$150.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
250 MG
$150.00
1 G
$436.00

About This Item

Empirical Formula (Hill Notation):
C16H10N2O8
CAS Number:
Molecular Weight:
358.26
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$150.00


0 left in stock

Request a Bulk Order

Assay

≥97%

SMILES string

N(=N\c2cc(cc(c2)C(=O)O)C(=O)O)/c1cc(cc(c1)C(=O)O)C(=O)O

InChI

1S/C16H10N2O8/c19-13(20)7-1-8(14(21)22)4-11(3-7)17-18-12-5-9(15(23)24)2-10(6-12)16(25)26/h1-6H,(H,19,20)(H,21,22)(H,23,24)(H,25,26)/b18-17+

InChI key

MXBBZODCMBYDCL-ISLYRVAYSA-N

Legal Information

Product of SiKEMIA

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service