Skip to Content
Merck
All Photos(1)

Documents

SIK1102-10

Sigma-Aldrich

12,12,13,13,14,14,15,15,16,16,17,17,17-Tridecafluoroheptadecane-1-thiol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23F13S
CAS Number:
Molecular Weight:
506.41
UNSPSC Code:
12352105
NACRES:
NA.23

Assay

97%

form

solid

shelf life

3 years under the recommended conditions

color

white

Application

These products are used to form water-repellent surface coating and/or lubricated surfaces on coinage metal materials (such as gold, silver).
The fluorinated carbon gives omniphobic properties to the surface. The coated surface is more water repellent when a product having more fluorinated carbons is used.
The thiol group provide a linker that covalently bond to the coinage metal surface.
The long carbon chain length allows for easy formation of a self-assembled monolayer.

Legal Information

Product of SiKEMIA

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service