Skip to Content
Merck
All Photos(1)

Key Documents

S550

Sigma-Aldrich

Salicyloyl hydrazide

98%

Synonym(s):

Salicyl hydrazide, Salicylhydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CONHNH2
CAS Number:
Molecular Weight:
152.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

147-150 °C (lit.)

SMILES string

NNC(=O)c1ccccc1O

InChI

1S/C7H8N2O2/c8-9-7(11)5-3-1-2-4-6(5)10/h1-4,10H,8H2,(H,9,11)

InChI key

XSXYESVZDBAKKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laith Q Al-Mawsawi et al.
Bioorganic & medicinal chemistry letters, 17(23), 6472-6475 (2007-10-24)
The previously discovered salicylhydrazide class of compounds displayed potent HIV-1 integrase (IN) inhibitory activity. The development of this class of compounds as antiretroviral agents was halted due to cytotoxicity in the nanomolar to sub-micromolar range. We identified a novel class
Xuefei Cao et al.
Current cancer drug targets, 9(2), 189-201 (2009-03-12)
Previously, we described a series of salicylhydrazide compounds with potent anti-cancer activities against a panel of human cancer cell lines derived from different origins. Preclinical evaluation showing efficacy both in vitro and in vivo in human cancer models indicated that
Paul S Francis
Luminescence : the journal of biological and chemical luminescence, 19(4), 205-208 (2004-08-03)
The chemiluminescence accompanying the oxidation of salicylic hydrazide (2-hydroxybenzoic acid hydrazide) with hypochlorite, hypobromite, N-chlorosuccinimide, N-bromosuccinimide or hydrogen peroxide with cobalt(II) matched the photoluminescence emission of salicylic acid. In a related reaction, the oxidation of a mixture of isoniazid and
S A Thompson et al.
British journal of pharmacology, 142(1), 97-106 (2004-04-22)
1. A high-throughput assay utilizing the voltage/ion probe reader (VIPR) technology identified salicylidene salicylhydrazide (SCS) as being a potent selective inhibitor of alpha2beta1gamma1 GABA(A) receptors with a maximum inhibition of 56+/-5% and an IC(50) of 32 (23, 45) nm. 2.
Muhib Ahmed et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 32(4), 671-682 (2019-06-24)
Hydrazide ligand, (Z)-N'-(6-oxo-1,10-phenanthrolin-5(6H)-ylidene)isonicotinohydrazide, 1 forms from a 1:1 Schiff base condensation reaction between isoniazid (INH) and 1,10-phenanthroline-5,6-dione (phendione). Ag+ and Mn2+ complexes with 1:2 metal:ligand stoichiometry are prepared: [Ag(1)2]NO3, [Ag(1)2]BF4 and [Mn(1)2](NO3)2. Polymeric {[Ag(1)(NO3)]}n has 1:1 stoichiometry and forms upon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service