Skip to Content
Merck
All Photos(1)

Key Documents

QBD-AQ-11960LF

Sigma-Aldrich

AQuora®750-NHS Ester

Synonym(s):

2-((1E,3E,5E)-7-((E)-1-(31-((2,5-dioxopyrrolidin-1-yl)oxy)-3,31-dioxo-7,10,13,16,19,22,25,28-octaoxa-4-azahentriacontyl)-3-methyl-3-(15-oxo-2,5,8,11-tetraoxa-14-azaheptadecan-17-yl)-5-sulfonatoindolin-2-ylidene)hepta-1,3,5-trie

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$307.00

$307.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$307.00

About This Item

Empirical Formula (Hill Notation):
C81H126N5O32S2
Molecular Weight:
1746.01
UNSPSC Code:
12352108
NACRES:
NA.01

$307.00


Check Cart for Availability

Request a Bulk Order

Quality Level

SMILES string

CC(C1=C(N/2CCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCC(ON3C(CCC3=O)=O)=O)=O)C=CC(S(=O)([O-])=O)=C1)(CCC(NCCOCCOCCOCCOC)=O)C2=C/C=C/C=C/C=C/C4=[N+](CCOCCOCCOCCOC)C(C=CC(S(=O)([O-])=O)=C5)=C5C4(CCOCCOCCOCCOC)C

General description

AQuora® 750-NHS Ester is an amine-reactive fluorophore designed to improve solubility during labeling and of the dye labeled conjugate. As a result, the dye labeled conjugates made with AQuora® 750-NHS Ester yield enhanced signal and signal-to-noise ratios in fluorescence-based applications including fluorescent western blotting, fluorescence-based microscopy, flow cytometry, and cell-based assays.

This AQuora® 750-NHS Ester dye is activated with a N-hydroxysuccinimide (NHS) ester group which reacts with free primary amines, such as the amines on the side chain of lysines, forming a stable amide bond.

Legal Information

AQuora is a registered trademark of Quanta BioDesign, Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service