Skip to Content
Merck
All Photos(1)

Key Documents

QBD11305

Sigma-Aldrich

Amino-dPEG ®24-amido-dPEG ®24-acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$628.00
1000 MG
$2,360.00

$628.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$628.00
1000 MG
$2,360.00

About This Item

Empirical Formula (Hill Notation):
C102H204N2O51
Molecular Weight:
2274.69
UNSPSC Code:
12352106
NACRES:
NA.22

$628.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: carboxyl reactive

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(O)=O)=O

InChI

1S/C102H204N2O51/c103-3-7-110-11-15-114-19-23-118-27-31-122-35-39-126-43-47-130-51-55-134-59-63-138-67-71-142-75-79-146-83-87-150-91-95-154-99-97-152-93-89-148-85-81-144-77-73-140-69-65-136-61-57-132-53-49-128-45-41-124-37-33-120-29-25-116-21-17-112-13-9-

InChI key

FCQAQQLROKWRBB-UHFFFAOYSA-N

Features and Benefits

Amino-dPEG®₂₄-amido-dPEG®₂₄-acid can be used to introduce a hydrophilic dPEG through reaction with either the amine or the carboxyl.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service