Skip to Content
Merck
All Photos(1)

Key Documents

QBD10808

Sigma-Aldrich

Lipoamido-dPEG®12-acid

Synonym(s):

Polyethylene glycol, alpha-(2-carboxyethyl)-omega-[2-[[5-(3R)-1,2-dithiolan-3-yl-1-oxopentyl]amino]ethoxy]poly(oxy-1,2-ethanediyl)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$602.00
1000 MG
$2,510.00

$602.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$602.00
1000 MG
$2,510.00

About This Item

Empirical Formula (Hill Notation):
C35H67NO15S2
CAS Number:
Molecular Weight:
806.03
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$602.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations
reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: gold reactive

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCCCC1SSCC1)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(O)=O

InChI

1S/C35H67NO15S2/c37-34(4-2-1-3-33-6-32-52-53-33)36-7-9-41-11-13-43-15-17-45-19-21-47-23-25-49-27-29-51-31-30-50-28-26-48-24-22-46-20-18-44-16-14-42-12-10-40-8-5-35(38)39/h33H,1-32H2,(H,36,37)(H,38,39)

InChI key

KTDABPXYRAJNFQ-UHFFFAOYSA-N

Features and Benefits

Lipoamido-dPEG®12-acid permits crosslinking of a molecule with an amine functional group to a metal surface such as gold. The lipoamide group readily forms strong, stable dative bonds with metals such as gold, while the acid group reacts with amines. The dPEG® spacers are hydrophilic, non-immunogenic, single molecular weight compounds of exact length. Precise spacing control of the crosslinked molecules is possible because of this single molecular weight nature. The dPEG® spacers also improve the water solubility of the target molecules.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service