Skip to Content
Merck
All Photos(1)

Key Documents

QBD10644

Sigma-Aldrich

Lipoamido-dPEG®36-TFP ester

>90% (HPLC)

Synonym(s):

Lipoamido-PEG-TFP ester, Lipoamido-PEG36-TFP ester, TFP-PEG1500-Lipoamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$1,340.00

$1,340.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$1,340.00

About This Item

Empirical Formula (Hill Notation):
C89H163F4NO39S2
Molecular Weight:
2011.35
UNSPSC Code:
12352108
NACRES:
NA.22

$1,340.00


Estimated to ship on30 April 2025


Request a Bulk Order

Assay

>90% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

Lipoamido-dPEG36-TFP ester is a monodispersed, crosslinking PEGylation reagent that joins free amines to metal surfaces. The discrete PEG (dPEG) portion of this crosslinker is comparable to a PEG1500. The single molecular weight, discrete-chain-length polyethylene glycol (dPEG) product consists of alpha-lipoic acid and a 2,3,5,6-tetrafluorophenol (TFP) ester on opposite ends of a 120 atoms-long (136.1 Å) dPEG spacer. The lipoic acid group forms two (2) coordinate covalent bonds (dative bonds) with metal, especially gold and silver. The TFP ester reacts with free amines to form amide bonds. This product has been utilized for sensor development, peptide or protein immobilization on metal surfaces, and the formation of self-assembled monolayers (SAMs).

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service