Skip to Content
Merck
All Photos(3)

Key Documents

Q1506

Sigma-Aldrich

8-Quinolinesulfonyl chloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$75.60
10 G
$144.00

$75.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$75.60
10 G
$144.00

About This Item

Empirical Formula (Hill Notation):
C9H6ClNO2S
CAS Number:
Molecular Weight:
227.67
Beilstein:
156347
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$75.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

126-129 °C (lit.)

SMILES string

ClS(=O)(=O)c1cccc2cccnc12

InChI

1S/C9H6ClNO2S/c10-14(12,13)8-5-1-3-7-4-2-6-11-9(7)8/h1-6H

InChI key

JUYUYCIJACTHMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijay K Agrawal et al.
European journal of medicinal chemistry, 39(7), 593-600 (2004-07-09)
Quantitative structure-activity-relationship (QSAR) study on aromatic/heterocyclic sulfonamides containing 8-quinoline-sulfonyl carbonic anhydrase (CA) inhibitors has been carried out topologically using first-order valence connectivity index ((1)chi(v)). Excellent results are obtained against all the three isozymes; CA I, II and IV of the
J Borras et al.
Bioorganic & medicinal chemistry, 7(11), 2397-2406 (2000-01-13)
Reaction of 20 aromatic/heterocyclic sulfonamides containing a free amino, imino, hydrazino or hydroxyl group, with 8-quinoline-sulfonyl chloride afforded a series of water-soluble (as hydrochloride or triflate salts) compounds. The new derivatives were assayed as inhibitors of the zinc enzyme carbonic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service