Skip to Content
Merck
All Photos(3)

Key Documents

Q0750

Sigma-Aldrich

Quinidine hydrochloride monohydrate

technical grade

Synonym(s):

6′-Methoxycinchonan-9-ol hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$96.40
10 G
$139.00

$96.40


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$96.40
10 G
$139.00

About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · HCl · H2O
CAS Number:
Molecular Weight:
378.89
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$96.40


Estimated to ship on11 April 2025Details


Request a Bulk Order

grade

technical grade

impurities

≤20% Dihydroquinidine (according to USP specifications., actual content given on label)

SMILES string

O.Cl.[H][C@@]1(C[C@@H]2CC[N@H]1C[C@@H]2C=C)[C@@H](O)c3ccnc4ccc(OC)cc34

InChI

1S/C20H24N2O2.ClH.H2O/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;1H;1H2/t13-,14-,19+,20-;;/m0../s1

InChI key

SGVZDMWHXVXUBY-KAIFKDDSSA-N

Application

Studied for pharmacological activity causing energy shifts from mitochondrial respiration to glycolysis

Biochem/physiol Actions

Class IA antiarrhythmic; potassium channel blocker.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Fukao et al.
European journal of drug metabolism and pharmacokinetics, 41(6), 819-824 (2015-12-03)
We have recently found an H We investigated the uptake and efflux of quinidine in MDCK cells. The uptake of 100 µM quinidine into MDCK cells was decreased by acidification of extracellular pH or alkalization of intracellular pH. In addition, the
Keisuke Oda et al.
The Journal of pharmacy and pharmacology, 69(12), 1736-1744 (2017-10-06)
Possible interaction of green tea beverage (GT) containing cyclodextrins and high concentration catechins, a drinking water, with P-glycoprotein (P-gp) substrates was examined in vitro and in vivo. Effects of GT on the uptake of rhodamine 123 by LLC-GA5-COL150 cells and intestinal efflux
Faisal E Al Jofi et al.
Cytotherapy, 20(5), 650-659 (2018-03-21)
Compelling evidence indicates that metformin, a low-cost and safe orally administered biguanide prescribed to millions of type 2 diabetics worldwide, induces the osteoblastic differentiation of mesenchymal stromal cells (MSCs) through the 5' adenosine monophosphate (AMP)-activated protein kinase (AMPK) pathway. As
Rajesh Chavan et al.
Xenobiotica; the fate of foreign compounds in biological systems, 51(3), 251-261 (2020-10-21)
Nafithromycin is a next generation lactone ketolide antibiotic slated to enter phase III clinical development in India for the treatment of CABP as a shorter 800 mg-OD X3 day therapeutic regimen. Nafithromycin exhibits potent activity against MDR Streptococcus pneumoniae including erythromycin and
Yingshan Han et al.
Journal of medical virology, 91(7), 1182-1190 (2019-02-26)
Studies aimed at repurposing existing drugs revealed that some antimalarial compounds possess anti-Zika virus (anti-ZIKV) activity. Here, we further tested 14 additional antimalarial drugs and their metabolites or analogs for anti-ZIKV activity using a phenotypic screening approach. We identified four

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service