Skip to Content
Merck
All Photos(2)

Key Documents

P65606

Sigma-Aldrich

2-Pyridylacetic acid hydrochloride

99%

Synonym(s):

2-Pyridineacetic acid hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$91.00
25 G
$146.00

$91.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$91.00
25 G
$146.00

About This Item

Empirical Formula (Hill Notation):
C7H7NO2 · HCl
CAS Number:
Molecular Weight:
173.60
Beilstein:
3696607
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$91.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

powder

mp

135 °C (dec.) (lit.)

SMILES string

Cl[H].OC(=O)Cc1ccccn1

InChI

1S/C7H7NO2.ClH/c9-7(10)5-6-3-1-2-4-8-6;/h1-4H,5H2,(H,9,10);1H

InChI key

MQVISALTZUNQSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service