Skip to Content
Merck
All Photos(2)

Key Documents

P57409

Sigma-Aldrich

2,2′-Pyridil

97%

Synonym(s):

Di-2-pyridylglyoxal

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$92.60
25 G
$321.00

$92.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$92.60
25 G
$321.00

About This Item

Empirical Formula (Hill Notation):
C12H8N2O2
CAS Number:
Molecular Weight:
212.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$92.60


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

154-156 °C (lit.)

SMILES string

O=C(C(=O)c1ccccn1)c2ccccn2

InChI

1S/C12H8N2O2/c15-11(9-5-1-3-7-13-9)12(16)10-6-2-4-8-14-10/h1-8H

InChI key

PIINXYKJQGMIOZ-UHFFFAOYSA-N

Gene Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farheen Fatima Khan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(41), 9737-9746 (2019-05-28)
In the recent years, there has been an emerging research interest in the domain of C-C bond-cleavage reactions. The present contribution deals with the redox-mediated dioxygen activation and C-C bond cleavage in a diruthenium complex [(acac)2 RuII (μ-L1)RuII (acac)2 ]
Rukhsana Anjum et al.
Inorganic chemistry, 58(20), 13709-13723 (2019-07-25)
A series of eight bis(thiosemicarbazone) ligands and 16 of their respective copper(II) and zinc(II) complexes containing a combination of hydrogen, methyl, pyridyl, phenyl, and/or ethyl substituents at the diimine position of the ligand backbone were synthesized and characterized. The objective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service