Skip to Content
Merck
All Photos(1)

Key Documents

P49503

Sigma-Aldrich

Piperonylamine

97%

Synonym(s):

1,3-Benzodioxole-5-methylamine, 3,4-(Methylenedioxy)benzylamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$172.00

$172.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$172.00

About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
136996
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$172.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.564 (lit.)

bp

138-139 °C/13 mmHg (lit.)

density

1.214 g/mL at 25 °C (lit.)

SMILES string

NCc1ccc2OCOc2c1

InChI

1S/C8H9NO2/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-3H,4-5,9H2

InChI key

ZILSBZLQGRBMOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariana C F C B Damião et al.
Archiv der Pharmazie, 347(12), 885-895 (2014-10-07)
A novel class of benzo[d][1,3]dioxol-5-ylmethyl alkyl/aryl amide and ester analogues of capsaicin were designed, synthesized, and evaluated for their cytotoxic activity against human and murine cancer cell lines (B16F10, SK-MEL-28, NCI-H1299, NCI-H460, SK-BR-3, and MDA-MB-231) and human lung fibroblasts (MRC-5).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service