Skip to Content
Merck
All Photos(3)

Key Documents

P43602

Sigma-Aldrich

3-(Chloromethyl)pyridine hydrochloride

96%

Synonym(s):

3-Picolyl chloride hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$57.20
25 G
$163.00

$57.20


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$57.20
25 G
$163.00

About This Item

Empirical Formula (Hill Notation):
C6H6ClN · HCl
CAS Number:
Molecular Weight:
164.03
Beilstein:
3689156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$57.20


0 left in stock

Request a Bulk Order

Assay

96%

form

solid

mp

137-143 °C (lit.)

SMILES string

Cl[H].ClCc1cccnc1

InChI

1S/C6H6ClN.ClH/c7-4-6-2-1-3-8-5-6;/h1-3,5H,4H2;1H

InChI key

UZGLOGCJCWBBIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A F McFee et al.
Environmental and molecular mutagenesis, 14(4), 207-220 (1989-01-01)
Three pairs of structurally similar carcinogenic/non-carcinogenic chemicals were tested for in vivo genotoxic activity in B6C3F1 mice. The carcinogenic/non-carcinogenic pairs, respectively, were o-toluidine hydrochloride/o-anthranilic acid, 4-chloro-o-phenylenediamine/4-nitro-o-phenylenediamine, and 3-(chloromethyl)pyridine hydrochloride/2-(chloromethyl)pyridine hydrochloride. Bone marrow cells from mice given intraperitoneal injections of up
D B McGregor et al.
Environmental mutagenesis, 9(2), 143-160 (1987-01-01)
Nine substances were tested for their mutagenic potential in the L5178Y tk+/tk- mouse lymphoma cell forward mutation assay, by means of procedures based upon those described by Clive and Spector (Mutat Res 44:269-278, 1975) and Clive et al (Mutat Res

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service