Skip to Content
Merck
All Photos(1)

Key Documents

P40506

Sigma-Aldrich

N-Phthaloylglycine

97%

Synonym(s):

1,3-Dioxo-2-isoindolineacetic acid, Phthalimidoacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$80.80
100 G
$279.00

$80.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$80.80
100 G
$279.00

About This Item

Empirical Formula (Hill Notation):
C10H7NO4
CAS Number:
Molecular Weight:
205.17
Beilstein:
184174
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

$80.80


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

193-196 °C (lit.)

SMILES string

OC(=O)CN1C(=O)c2ccccc2C1=O

InChI

1S/C10H7NO4/c12-8(13)5-11-9(14)6-3-1-2-4-7(6)10(11)15/h1-4H,5H2,(H,12,13)

InChI key

WQINSVOOIJDOLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O abu Salach et al.
Pharmaceutical research, 11(10), 1429-1434 (1994-10-01)
Glycine, in addition to GABA, is one of the most important neurotransmitter amino acids. The described structure pharmacokinetic pharmacodynamic relationships (SPPR) study explored the possibility of utilizing phthaloyl derivatives of glycine as new antiepileptics. This was carried out by investigating
Nasima Arshad et al.
Journal of photochemistry and photobiology. B, Biology, 117, 228-239 (2012-11-20)
Ni(II), Cu(II) and Zn(II) metal complexes of N-phthaloylglycine were synthesized, characterized, reported for single crystal X-ray diffraction analysis for Ni(II) complex, and investigated for their binding with DNA under physiological conditions, using spectroscopic (UV-visible and fluorescence) and hydrodynamic techniques. Experimental
Julija Matijević-Sosa et al.
Acta pharmaceutica (Zagreb, Croatia), 58(2), 231-236 (2008-06-03)
The aim of this study was to investigate the activity of N-phthaloyl-glycine-hydroxamic acid (Phth-Gly-HA) as a new iron chelator in vivo to be used in iron overload diseases. After intraperitoneal application of Phth-Gly-HA to male rats (1 mg kg(-1) body

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service