Skip to Content
Merck
All Photos(1)

Key Documents

P36401

Sigma-Aldrich

3-Phenyltoluene

95%

Synonym(s):

3-Methylbiphenyl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$89.80

$89.80


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 G
$89.80

About This Item

Linear Formula:
C6H5C6H4CH3
CAS Number:
Molecular Weight:
168.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$89.80


0 left in stock

Request a Bulk Order

Assay

95%

refractive index

n20/D 1.602 (lit.)

bp

272 °C (lit.)

mp

4-5 °C (lit.)

density

1.018 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1)-c2ccccc2

InChI

1S/C13H12/c1-11-6-5-9-13(10-11)12-7-3-2-4-8-12/h2-10H,1H3

InChI key

NPDIDUXTRAITDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp Denk et al.
Analytical and bioanalytical chemistry, 410(14), 3395-3404 (2018-04-07)
Elastic therapeutic tapes are an important tool in the field of physical therapy and medicine. These tapes contain types of adhesive. However, sensory evaluations revealed the release of pronounced and irritating odors of the tapes. Negative odors were, amongst others

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service