P17008
L-Phenylalanine
99%
Synonym(s):
(S)-2-Amino-3-phenylpropionic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
165.19
Beilstein:
1910408
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Assay
99%
form
powder
optical activity
[α]25/D −32.7°, c = 2 in H2O
optical purity
ee: 98% (GLC)
color
white
mp
270-275 °C (dec.) (lit.)
SMILES string
N[C@@H](Cc1ccccc1)C(O)=O
InChI
1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1
InChI key
COLNVLDHVKWLRT-QMMMGPOBSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ann Mullally et al.
Blood, 121(18), 3692-3702 (2013-03-15)
Interferon-α (IFNα) is an effective treatment of patients with myeloproliferative neoplasms (MPNs). In addition to inducing hematological responses in most MPN patients, IFNα reduces the JAK2V617F allelic burden and can render the JAK2V617F mutant clone undetectable in some patients. The
Stefanie Keil et al.
Pediatrics, 131(6), e1881-e1888 (2013-05-22)
Sapropterin dihydrochloride, the synthetic form of 6R-tetrahydrobiopterin (BH4), is an approved drug for the treatment of patients with BH4-responsive phenylketonuria (PKU). The purpose of this study was to assess genotypes and data on the long-term effects of BH4/sapropterin on metabolic
Vimesh A Avlani et al.
American journal of physiology. Endocrinology and metabolism, 304(10), E1097-E1104 (2013-03-28)
In addition to its acute effects on hormone secretion, epithelial transport, and shape change, the calcium-sensing receptor (CaSR) modulates the expression of genes that control cell survival, proliferation, and differentiation as well as the synthesis of peptide hormones and enzymes.
John J Mitchell et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 13(8), 697-707 (2011-05-11)
Phenylalanine hydroxylase deficiency is an autosomal recessive disorder that results in intolerance to the dietary intake of the essential amino acid phenylalanine. It occurs in approximately 1:15,000 individuals. Deficiency of this enzyme produces a spectrum of disorders including classic phenylketonuria
Bart Pennings et al.
The American journal of clinical nutrition, 98(1), 121-128 (2013-05-03)
Older individuals generally experience a reduced food-chewing efficiency. As a consequence, food texture may represent an important factor that modulates dietary protein digestion and absorption kinetics and the subsequent postprandial protein balance. We assessed the effect of meat texture on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service