Skip to Content
Merck
All Photos(1)

Key Documents

N6885

Sigma-Aldrich

Sodium 2-naphthalenesulfonate

≥95% (HPLC)

Synonym(s):

2-Naphthalenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$44.80

$44.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$44.80

About This Item

Empirical Formula (Hill Notation):
C10H7NaO3S
CAS Number:
Molecular Weight:
230.22
Beilstein:
3659024
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$44.80


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

SMILES string

[Na+].[O-]S(=O)(=O)c1ccc2ccccc2c1

InChI

1S/C10H8O3S.Na/c11-14(12,13)10-6-5-8-3-1-2-4-9(8)7-10;/h1-7H,(H,11,12,13);/q;+1/p-1

InChI key

YWPOLRBWRRKLMW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elodie Alinat et al.
Electrophoresis, 36(14), 1555-1563 (2015-03-31)
This work focuses on the development of a CE method allowing, for the first time, the simultaneous separation of the underivatized first seven cellodextrin oligomers (glucose, cellobiose, cellotriose, cellotetraose, cellopentaose, cellohexaose, and celloheptaose), with a view to analyze the hydrolysates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service