Skip to Content
Merck
All Photos(1)

Key Documents

N3405

Sigma-Aldrich

2-Naphthoxyacetic acid

98%

Synonym(s):

BNOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH2CO2H
CAS Number:
Molecular Weight:
202.21
Beilstein:
1074148
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

powder

mp

151-154 °C (lit.)

SMILES string

OC(=O)COc1ccc2ccccc2c1

InChI

1S/C12H10O3/c13-12(14)8-15-11-6-5-9-3-1-2-4-10(9)7-11/h1-7H,8H2,(H,13,14)

InChI key

RZCJYMOBWVJQGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanghamitra Atta et al.
Journal of photochemistry and photobiology. B, Biology, 111, 39-49 (2012-04-20)
We report a novel technique for controlled release of plant growth regulators (PGRs) by sunlight using photoremovable protecting group (PRPG) as a delivery device. In the present work, carboxyl-containing PGRs of the auxin group [indoleacetic acid (IAA) and naphthoxyacetic acid
A Kokkinou et al.
Carbohydrate research, 332(1), 85-94 (2001-06-14)
The structure of the complex of beta-cyclodextrin (cyclomaltoheptaose) with beta-naphthyloxyacetic acid was studied in solid state by X-ray diffraction and in aqueous solution by 1H NMR spectroscopy. The complex crystallizes in the channel mode, space group C2, with a stoichiometry
Chun Zhou et al.
Environmental monitoring and assessment, 154(1-4), 233-239 (2008-06-17)
A reliable and sensitive competitive fluorescence immunoassay for the quantitative determination of naphthalene (NA) was developed. 2-naphthoxy acetic acid (NAA) was selected as the hapten of naphthalene. Active ester method (AEM) was used to couple the NAA to carrier proteins
L Li et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56A(8), 1513-1521 (2000-07-25)
A cyclodextrin induced room temperature phosphorimetry (CD-RTP) for determine beta-NOA, which using I- as a heavy atom perturber (HAP) and sodium sulfite as a deoxygenator, was developed. The phosphorescence peak wavelength maxima lambda(ex)/lambda(em) = 287/496,521 nm. The analytical curve of
M Donbrow et al.
The Journal of pharmacy and pharmacology, 34(4), 215-224 (1982-04-01)
Stability constants and thermodynamic parameters have been evaluated for the complexation reaction in aqueous solution of caffeine with beta-naphthoxy acetic acid. The values were higher than those previously reported for the complexation of other ligands with methyl xanthines. In nearly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service