N29805
1,9-Nonanedithiol
95%
Synonym(s):
1,9-Dimercaptononane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HS(CH2)9SH
CAS Number:
Molecular Weight:
192.39
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.4999 (lit.)
bp
284 °C (lit.)
density
0.952 g/mL at 25 °C (lit.)
SMILES string
SCCCCCCCCCS
InChI
1S/C9H20S2/c10-8-6-4-2-1-3-5-7-9-11/h10-11H,1-9H2
InChI key
GJRCLMJHPWCJEI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
215.6 °F - closed cup
Flash Point(C)
102 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Wei Zhao et al.
Small (Weinheim an der Bergstrasse, Germany), 11(35), 4509-4516 (2015-06-04)
The ability to tune gauge factors in terms of magnitude and orientation is important for wearable and conformal electronics. Herein, a sensor device is described which is fabricated by assembling and printing molecularly linked thin films of gold nanoparticles on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service