Skip to Content
Merck
All Photos(2)

Key Documents

N27004

Sigma-Aldrich

2-Nitrothiophene

85%, technical grade

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$84.20

$84.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$84.20

About This Item

Empirical Formula (Hill Notation):
C4H3NO2S
CAS Number:
Molecular Weight:
129.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$84.20


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Assay

85%

form

crystals

impurities

15% 3-nitrothiophene

bp

224-225 °C (lit.)

mp

43-45 °C (lit.)

SMILES string

[O-][N+](=O)c1cccs1

InChI

1S/C4H3NO2S/c6-5(7)4-2-1-3-8-4/h1-3H

InChI key

JIZRGGUCOQKGQD-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eachan O Johnson et al.
Nature, 571(7763), 72-78 (2019-06-21)
New antibiotics are needed to combat rising levels of resistance, with new Mycobacterium tuberculosis (Mtb) drugs having the highest priority. However, conventional whole-cell and biochemical antibiotic screens have failed. Here we develop a strategy termed PROSPECT (primary screening of strains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service