Skip to Content
Merck
All Photos(1)

Documents

N24005

Sigma-Aldrich

6-Nitroquinoline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2O2
CAS Number:
Molecular Weight:
174.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

151-153 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc2ncccc2c1

InChI

1S/C9H6N2O2/c12-11(13)8-3-4-9-7(6-8)2-1-5-10-9/h1-6H

InChI key

SMHPLBXIVNQFBA-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fanny Widjaja et al.
Journal of the American Society for Mass Spectrometry, 28(7), 1392-1405 (2017-05-01)
The gas-phase reactivity of two aromatic carbon-centered σ,σ-biradicals (meta-benzyne analogs) and a related monoradical towards small oligonucleotides of differing lengths was investigated in a Fourier-transform ion cyclotron resonance (FT-ICR) mass spectrometer coupled with laser-induced acoustic desorption (LIAD). The mono- and
Xiulong Shen et al.
Chemical research in toxicology, 32(3), 348-361 (2019-03-01)
Enzymatic one-electron reduction of heterocyclic N-oxides can lead to the intracellular generation of reactive oxygen species via several different chemical pathways. These reactions may be relevant to hypoxia-selective anticancer drugs, antimicrobial agents, and unwanted toxicity of heterocylic nitrogen compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service