N12104
4-Nitro-2,1,3-benzothiadiazole
99%
Synonym(s):
4-Nitropiazthiole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H3N3O2S
CAS Number:
Molecular Weight:
181.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
solid
mp
108-110 °C (lit.)
SMILES string
[O-][N+](=O)c1cccc2nsnc12
InChI
1S/C6H3N3O2S/c10-9(11)5-3-1-2-4-6(5)8-12-7-4/h1-3H
InChI key
IWQKAMJGVIHECB-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shi-Wen Lv et al.
Journal of hazardous materials, 387, 122011-122011 (2020-01-14)
Nowadays, designing highly active photocatalysts for pollutant degradation under visible light still remains a challenging problem. Herein, a novel benzothiadiazole functionalized Co-doped MOF-based photocatalyst with electron deficient unit was first synthesized via a feasible step-by-step assembly strategy. Benzothiadiazole, as typical
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service