MT51343160
Mettler-Toledo InLab® Micro & Micro Pro pH electrode
Micro without integrated temperature sensor, S7 connector, L × diam. 60 mm × 3 mm
Synonym(s):
narrow pH electrode, vial electrode
About This Item
Recommended Products
material
glass shaft
glass, U membrane
description
Reference electrolyte 3 mol/L KCl
Reference system ARGENTHAL™ with Ag+ -trap
Without cable
feature
S7 connector
manufacturer/tradename
Mettler-Toledo 51343160
temperature measuring range
0-80 °C
L × diam.
60 mm × 3 mm
pH range
0-14
Looking for similar products? Visit Product Comparison Guide
General description
Ideal for measuring in very small vessels such as vials or cuvettes.
Designed with a shaft diameter of just 3 mm the InLab Micro is able to measure pH in the smallest vessels used in the laboratory.
Combination design avoids use of separate reference and indicating electrodes. The innovative Safelock closing ring gives easy access for re-filling reference electrolyte and complete sealing when closed. Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
- 3 mm diameter shaft
- Combination design
- Safelock closing ring
- Detachable cable
Features and Benefits
Legal Information
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service