MT51343070
Mettler-Toledo InLab® Science & Science Pro pH electrode
S7 connector, L × diam. 120 mm × 12 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
glass membrane (A41)
glass shaft
description
Reference electrolyte 3 mol/L KCl
Reference system ARGENTHAL™ with Ag+ -trap
manufacturer/tradename
Mettler-Toledo 51343161
temperature measuring range
0-100 °C
L × diam.
120 mm × 12 mm
pH range
0-12
Looking for similar products? Visit Product Comparison Guide
General description
InLab® Sience and Sience Pro are glass-body, combination pH electrode with S7 screw head and ground glass sleeve ideal for slurries and emulsions.
The sleeve junction increases flow rate and improves sample contact and can be easily cleaned.
The ARGENTHAL™ reference system improves service life and measurement accuracy at differing sample temperatures.
The sleeve junction increases flow rate and improves sample contact and can be easily cleaned.
The ARGENTHAL™ reference system improves service life and measurement accuracy at differing sample temperatures.
- ARGENTHAL reference
- Sleeve junction
- detachable cable
Science
Legal Information
ARGENTHAL is a trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service